2-(3-Hexyl-4-oxothiazolidin-2-ylidene)malononitrile ≥97%

Supplier: Ambeed
Danger

A1164010-1G A1164010-250MG A1164010-5G
77620-326EA 70.04 USD
77620-326 77866-165 77866-166
2-(3-Hexyl-4-oxothiazolidin-2-ylidene)malononitrile ≥97%
2-(3-Hexyl-4-oxothiazolidin-2-ylidene)malononitrile

Caution: for Research Use Only

Formula: C₁₂H₁₅N₃OS
CAS Number: 2084812-77-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR