Ascomycin ≥96%

Supplier: Ambeed
Danger

Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

A113253-100MG A113253-10MG A113253-50MG A113253-250MG
77599-634EA 47.15 USD
77599-634 77599-636 77623-940 77876-843
Ascomycin ≥96%
Ascomycin

Caution: for Research Use Only

Formula: C₄₃H₆₉NO₁₂
MW: 790 g/mol
MDL Number: MFCD06198665
CAS Number: 104987-12-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR