3-Amino-4-(trifluoromethyl)pyridine ≥95%
Supplier: Ambeed
|
Warning
|
A112272-5G
A112272-1G
A112272-250MG
77570-680EA
66
USD
77570-680
77610-890
77684-998
3-Amino-4-(trifluoromethyl)pyridine ≥95%
3-Amino-4-(trifluoromethyl)pyridine
Caution: for Research Use Only
Formula:
C₆H₅F₃N₂ |
MDL Number:
MFCD00173960 CAS Number: 175204-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...