EZ Rich Defined Medium Kit, Teknova
Supplier: Teknova
EZ Rich
M2105 M2105
200059-658EA 505.1 USD
CA200059-658 200059-658
EZ Rich Defined Medium Kit, Teknova
Media for Microbiology Prepared Media for Microbiology
Designed for the growth of E
MOPS mixture, ACGU solution, and EZ supplement should be stored frozen. The potassium phosphate dibasic solution and 20% glucose solution should be stored at room temperature.
coli and other enterobacteria. Kit consists of 100mL 10X MOPS mixture (200059-678), 50mL 0.132M potassium phosphate diabase solution (200059-652), 100mL 10X ACGU solution (200059-654), 200mL 5X supplement EZ (200059-656), and 100mL 20% glucose solution (100216-728). Kit makes 5 liters.