Septihol® Ethyl WFI st Alcohol Solution

Supplier: Steris
1S1108 1S1177 1S1196
77531-084CS 588 USD
77531-084 77531-086 77531-082
Septihol® Ethyl WFI st Alcohol Solution
Cleaning Agents Disinfectants
Septihol® Ethyl WFI st alcohol solution provides microbial control and removes residue from hard surfaces. It can be used straight out of the container and be applied poured or sprayed. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.

Septihol® Ethyl WFI st is a sterile mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI) designed to remove organic and inorganic residues in aseptic processing areas. Its EPA biocidal and fungicidal claims are proven in presence of 5% organic soil on hard, nonporous surfaces. Septihol Ethyl WFI st is double bagged and gamma irradiated for use in aseptic areas. The formula does not contain stabilizers or denaturants, so it evaporates quickly and completely.

CAS: 64-17-5; 67-63-0; 67-56-1
UN 1170
Hazard class: 3
Packing group: II

Caution: Danger, GHS02; GHS07; GHS08.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR