T-Clean Chamber Shine for Chamber Cleaning
Supplier: Tuttnauer
SH0006
77574-046CS 151.09 USD
77574-046
T-Clean Chamber Shine for Chamber Cleaning
Cleaning Agents Detergents and Surface Cleansers
T-Clean Chamber Shine Autoclave Cleaning Treatment Foam Spray is a safe and efficient revitalizing solution that restores the original finish to autoclave walls and stainless steel carts and trays.
  • Eliminates noxious odors
  • Non corrosive
  • Leaves no residue
Removes rust, spots, stains and corrosion, as well as scale, alkaline and mineral deposits.