Anti-CD247 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))

Supplier: Sino Biological
13142MM06P100 13142MM06P25
77444-246EA 280 USD
77444-246 77444-248
Anti-CD247 Mouse Monoclonal Antibody [clone: 06] (PE (Phycoerythrin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD3 zeta/CD247 (rh CD3 zeta/CD247; Catalog#13142-H09E; P20963-1; Arg52-Arg164) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 06
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR