Triolein 60%

Supplier: Ambeed
Warning

A744527-100G A744527-1000G A744527-500G
77569-406EA 5 USD
77569-406 77873-178 77873-179
Triolein 60%
Triolein

Caution: For research use only.

Formula: C₅₇H₁₀₄O₆
MDL Number: MFCD00137563
CAS Number: 122-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR