H-DAB(boc)-OH 95%

Supplier: Ambeed
Warning

A696341-5G A696341-1G A696341-250MG A696341-25G
77569-326EA 42 USD
77569-326 77595-072 77602-766 77880-444
H-DAB(boc)-OH 95%
H-DAB(boc)-OH

Caution: For research use only.

Formula: C₉H₁₈N₂O₄
MDL Number: MFCD00236842
CAS Number: 10270-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR