3-Amino-5-chlorobenzonitrile 96%

Supplier: Ambeed
Warning

A605903-1G A605903-5G
77569-156EA 18 USD
77569-156 77669-618
3-Amino-5-chlorobenzonitrile 96%
3-Amino-5-chlorobenzonitrile

Caution: For research use only.

Formula: C₇H₅ClN₂
MDL Number: MFCD06797224
CAS Number: 53312-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR