2-Fluoro-5-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Danger

A505290-25G A505290-5G A505290-1G
77568-982EA 313 USD
77568-982 77613-594 77673-322
2-Fluoro-5-(trifluoromethoxy)aniline 98%
2-Fluoro-5-(trifluoromethoxy)aniline

Caution: For research use only.

Formula: C₇H₅F₄NO
MDL Number: MFCD09832254
CAS Number: 116369-23-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR