Dimethyl methoxymethylenemalonate 95%

Supplier: Ambeed
Warning

A225653-500G A225653-25G A225653-1G
77568-180EA 454 USD
77568-180 77879-369 77879-368
Dimethyl methoxymethylenemalonate 95%
Dimethyl methoxymethylenemalonate

Caution: For research use only.

Formula: C₇H₁₀O₅
MDL Number: MFCD00038225
CAS Number: 22398-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR