Trimethyl phosphate 99.5%
Supplier: Ambeed
|
Danger
|
Synonyms:
Trimethylphosphate
A159403-100G
A159403-25G
A159403-500G
77567-316EA
14
USD
77567-316
77567-318
77871-796
Trimethyl phosphate 99.5%
Trimethyl phosphate
Caution: For research use only.
Formula:
C₃H₉O₄P MW: 140.08 g/mol Boiling Pt: 197.2 Melting Pt: -46 |
MDL Number:
MFCD00008348 CAS Number: 512-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...