Polyethyleneimine, M.W. 25000

Supplier: Ambeed
Danger

A1443158-25G A1443158-500G
77566-910EA 17 USD
77566-910 77874-847
Polyethyleneimine, M.W. 25000
Polyethyleneimine

Caution: For research use only.

Formula: (CH2CH2NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR