Carboxymethyl cellulose sodium salt, Viscosity 800-1200mpa.s
Supplier: Ambeed
|
Warning
|
Synonyms:
Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
A1361317-100G
A1361317-25G
A1361317-1000G
A1361317-500G
77566-698EA
11
USD
77566-698
77566-700
77592-568
77621-752
Carboxymethyl cellulose sodium salt, Viscosity 800-1200mpa.s
Carboxymethyl cellulose sodium salt
Caution: For research use only.
Formula:
[C₆H₇O₂(OH)x(OCH₂COONa)y]n |
MDL Number:
MFCD00081472 CAS Number: 9004-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...