Water, CellPro™, Ultrapure, DEPC treated
Supplier: Alkali Scientific
DC42236-CS
DC42110
DC42114
DC42-20L
DC422-4L
104027-054EA
527.68
USD
104027-054
104028-230
104027-418
104027-878
104027-136
Water, CellPro™, Ultrapure, DEPC treated
Water
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 UN: 3248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...