CellPro™ Agar Biological Grade
Supplier: Alkali Scientific
DC42238 DC42239 DC42240 DC42241
104027-726EA 49.22 USD
104027-726 104029-032 104027-800 104027-406
CellPro™ Agar Biological Grade
Media for Microbiology Prepared Media for Microbiology
Prepare high-quality agar plates with CellPro™ Agar Biological Grade, ensuring optimal conditions for microbial growth and analysis.
  • Suitable for many bacteriological applications
  • Broad technical specifications
  • Not recommended for growth of fastidious organisms
CellPro™ Agar Biological Grade is a high-quality agar medium designed for microbiological and molecular biology applications in laboratory settings. This agar powder is derived from select seaweed sources and undergoes stringent quality control processes to ensure purity and consistency. Biological Grade Agar is commonly used to prepare solid culture media for the cultivation and isolation of microorganisms, including bacteria, yeast, and fungi. It provides a solid substrate for microbial growth and is suitable for a wide range of microbiological techniques, such as colony counting, streaking and microbial identification.