CellPro™ LB + Chloramphenicol
Supplier: Alkali Scientific
DC0100 DC0101 DC0102 DC0096
104027-940EA 70.75 USD
104027-940 104027-394 104028-924 104028-564
CellPro™ LB + Chloramphenicol
Media for Microbiology Prepared Media for Microbiology
Support selective bacterial growth with CellPro™ LB + Chloramphenicol, providing a suitable medium for antibiotic-resistant strains.
  • Enhance selection with a potent combination of LB broth and chloramphenicol
  • Ensure efficient bacterial growth and selection for your experiments
  • Rely on consistent performance for reliable research outcomes
CellPro™ LB + Chloramphenicol is a nutrient-rich agar medium supplemented with chloramphenicol, an antibiotic that inhibits the growth of chloramphenicol-sensitive bacterial strains. This selective medium is commonly used in microbiology and molecular biology laboratories for the cultivation of bacteria harboring chloramphenicol resistance genes. By incorporating chloramphenicol into the LB agar, this medium facilitates the isolation and propagation of genetically modified bacterial cells in a controlled and selective environment.