CellPro™ LB (Lennox) + Ampicillin
Supplier: Alkali Scientific
DC0139 DC0144 DC0145 DC0060 DC0076 DC0056 DC0077 DC0078 DC0079 DC0080 DC0150
104027-206EA 64.6 USD
104027-206 104027-212 104028-664 104026-724 104028-968 104027-252 104027-008 104028-244 104027-644 104028-268 104027-630
CellPro™ LB (Lennox) + Ampicillin
Media for Microbiology Prepared Media for Microbiology
Support bacterial growth while selecting for ampicillin resistance with CellPro™ LB (Lennox) + Ampicillin, a specialized medium for molecular biology experiments.
  • Optimize selection with a powerful blend of Lennox LB broth and ampicillin
  • Maximize efficiency in bacterial transformation and plasmid amplification
  • Count on consistent results for confident discoveries
CellPro™ LB (Lennox) + Ampicillin is a modified variant of the classic Luria-Bertani (LB) agar, fortified with ampicillin to support the growth of ampicillin-resistant bacterial strains. This medium is commonly used in molecular biology laboratories for the cultivation and selection of transformed bacterial cells carrying ampicillin resistance genes. By incorporating ampicillin into the LB agar, this medium enables the isolation and propagation of genetically engineered bacteria in a controlled and selective manner.