CellPro™ LB + Ampicillin
Supplier: Alkali Scientific
DC0140 DC0143 DC0146 DC0147 DC0148 DC3560 DC0057 DC0058 DC0059 DC0061 DC0062 DC0063 DC0064 DC0065 DC0066 DC0067 DC0068 DC0069 DC0070 DC0071 DC0072 DC0073 DC0074 DC0047 DC0048 DC0050 DC0051 DC0052 DC0053 DC0054 DC0055 DC0149
104028-042EA 90.75 USD
104028-042 104028-542 104026-766 104028-520 104028-412 104027-062 104027-894 104028-890 104027-306 104028-964 104026-776 104029-040 104027-324 104028-354 104027-016 104027-792 104028-874 104028-252 104028-118 104027-320 104028-266 104027-736 104028-120 104028-360 104028-756 104027-144 104026-874 104028-574 104028-576 104027-796 104026-660 104026-870
CellPro™ LB + Ampicillin
Media for Microbiology Prepared Media for Microbiology
Prepare LB media with ampicillin supplementation using CellPro™ LB + Ampicillin, ensuring efficient growth of ampicillin-resistant bacterial strains.
  • Turbocharge cell culture with CellPro™ LB + Ampicillin
  • Optimal selection for antibiotic-resistant colonies
  • Superior bacterial transformation and protein expression
  • Reliable and consistent results, every time
  • Trusted by top researchers globally
CellPro™ LB + Ampicillin is a specialized bacterial growth medium designed for the cultivation of ampicillin-resistant bacterial strains. This nutrient-rich agar contains ampicillin, an antibiotic that inhibits the growth of bacteria lacking ampicillin resistance genes. Ideal for molecular biology and microbiology applications, this medium promotes the selective growth of genetically modified bacteria harboring ampicillin resistance plasmids, facilitating the isolation and maintenance of recombinant DNA constructs.