Femto HRP Chemiluminescent Substrate Kits
Supplier: Alkali Scientific
Optimize your chemiluminescent assays with the Femto HRP Chemiluminescent Substrate Kits, providing sensitive detection for various applications.
- Suitable detection for picogram levels of protein
- Sensitive and long lasting luminol-based
Femto HRP Chemiluminescent Substrate Kits offer a range of sensitivities for precise detection of low-abundance proteins in Western blotting. Following a simple protocol involving antibody incubation and substrate application, these kits deliver reliable results with minimal background noise. With detailed instructions and compatibility with various imaging systems, including CCD cameras, they facilitate efficient analysis of signal intensities for accurate protein quantification.