Square Media Bottles
Supplier: Alkali Scientific
VB1000 VB125 VB250 VB500
104028-210EA 112.29 USD
104028-210 104027-572 104027-548 104027-332
Square Media Bottles
Bottles Cell Culture Bottles
Store cell culture media securely with CellPro™ Square Media Bottles, designed for convenient storage and retrieval.
  • Molded-in graduation scales
  • Shrink-wrap tray modules
  • Non-pyrogenic, non-cytotoxic, and non-hemolytic
  • DNase/RNase-free and human DNA-free
CellPro™ Square Media Bottles are meticulously designed to meet the demanding needs of modern laboratories. Crafted with premium materials and innovative engineering, these bottles offer superior durability and functionality. The square shape not only maximizes storage efficiency but also ensures stability on shelves and during transportation. Each bottle features a secure screw cap closure to prevent leakage and contamination, providing optimal protection for valuable media solutions. With clear, easy-to-read volume graduations, users can accurately measure and dispense liquids with confidence. Whether storing cell culture media, reagents, or other valuable solutions, CellPro™ Square Media Bottles are the ideal choice for maintaining sample integrity and laboratory efficiency.