Flat Bottom Cell Culture Plates with Lids, Non-Treated, Sterile
Supplier: Alkali Scientific
TPN1006-NT TPN1012-NT TPN1024-NT TPN1048-NT TPN1096-NT
104028-632EA 110.75 USD
104028-632 104027-448 104026-710 104027-122 104027-584
Flat Bottom Cell Culture Plates with Lids, Non-Treated, Sterile
Microplates Cell Culture Plates
Facilitate cell culture experiments with flat bottom cell culture plates, featuring non-treated surfaces for versatility, supplied sterile with lids for ease of use in laboratory applications.
  • High clarity, 100% virgin polystyrene
  • Vacuum gas plasma tissue culture treatment
  • Clear lot number for batch tractability
  • Peel-to-open, individually wrapped and sterilized by gamma irradiation
  • Markings of well coordinates available for 12, 24, 48 and 96 well plates
  • Non-pyrogenic, DNase/RNase free
  • Gamma sterilized
Experience reliable and consistent cell culture results with flat bottom cell culture plates with lids. These plates, designed for versatility and convenience, feature a flat bottom and come with secure lids to maintain sterility throughout your experiments. Manufactured to the highest standards of quality, these plates are non-treated and sterile, providing a blank canvas for a variety of cell culture applications. Whether you're performing cell growth assays, proliferation studies, or other cell-based assays, these plates offer a dependable platform for your research needs.