Low Adhesion Cell Culture Plates for Suspension Cultures
Supplier: Alkali Scientific
TL41 TL42 TL43 TL44 TL45 TL46 TL47
104028-892EA 109.22 USD
104028-892 104027-588 104026-968 104027-808 104026-610 104028-526 104027-554
Low Adhesion Cell Culture Plates for Suspension Cultures
Microplates Cell Culture Plates
Facilitate suspension cultures with low adhesion cell culture plates, designed to minimize cell attachment and promote cell suspension.
  • Flat bottom
  • Sterilized by E-beam
  • Non treated surface allows for suspension cell cultures
  • Clear lot number for batch traceability
  • Peel-to-open, individually wrapped sterile
  • Markings of well coordinates available for 12, 24, 48 96 and 384 well plates
  • Non-pyrogenic, DNase/RNase Free
Low adhesion cell culture plates tailored for suspension cultures offers researchers a specialized tool for culturing cells in a suspended state. These plates feature a low-adhesion surface that minimizes cell attachment, allowing suspension cells to grow and proliferate freely. With this unique design, researchers can maintain cell viability and integrity while preventing cell aggregation. Ideal for studying cell signaling pathways, drug screening, and other suspension culture applications, low adhesion cell culture plates provide researchers with a reliable platform for their experiments.