Premium Cell Culture Lab Dishes
Supplier: Alkali Scientific
TDN0035 TDN0060 TDN0100 TDN0101 TDN0103 TDN0150
104028-506EA 161.46 USD
104028-506 104027-936 104026-936 104028-032 104028-134 104026-572
Premium Cell Culture Lab Dishes
Culture Dishes Cell Culture Dishes
Conduct cell culture experiments with precision using CellPro™ Premium Cell Culture Lab Dish, designed for optimal cell growth and experimentation.
  • Optically clear for microscopy applications
  • Large pronounced gripping rings
  • Air exchange vents
  • Pyrogen-free
  • Gamma sterilized
  • Tissue culture treated
  • Unique lid inner design for effective gas exchange
  • External grip for better handling
  • Excellent stackability
  • USP guideline compliant (USP class VI tested)
  • Non – pyrogenic, non – cytotoxic, DNase/RNase-free, and human DNA-free
The CellPro™ Cell Culture DishMore Product Information