SmartGlow™ Safe Green Pre Nucleic Acid Stains
Supplier: Alkali Scientific
Ensure safe and efficient nucleic acid staining with SmartGlow™ Safe Green Pre Nucleic Acid Stain, designed for enhanced visualization and safety.
- Replaces hazardous Ethidium bromide
- Excitation by UV or blue light
- Non-mutagenic, environmentally safe
- Two types: PS (Pre-Stain) LD (Loading Dye)
The SmartGlow™ Safe Green Pre Nucleic Acid Stain by CellPro™ is a cutting-edge solution for nucleic acid detection in molecular biology research. This innovative stain offers researchers a safe, sensitive, and environmentally friendly alternative to traditional nucleic acid dyes, minimizing hazardous waste and ensuring user safety without compromising performance. With its high sensitivity and low background fluorescence, SmartGlow™ enables accurate visualization and quantification of DNA and RNA in agarose gels, facilitating precise analysis of nucleic acid samples. Whether used for routine agarose gel electrophoresis or advanced molecular biology applications, SmartGlow™ Safe Green Pre Nucleic Acid Stain empowers researchers with confidence in their experimental results, driving discovery and innovation in the field of molecular biology.