CellPro™ Sodium Acetate, Molecular Biology Grade
Supplier: Alkali Scientific
DS0194 DB0261
104026-806EA 49.22 USD
104026-806 104027-646
CellPro™ Sodium Acetate, Molecular Biology Grade
Buffers Cloning Buffers
Facilitate nucleic acid precipitation with CellPro™ Sodium Acetate, ensuring molecular biology grade purity and reliability.
  • DNase, RNase, and Protease tested
  • Sterile
  • Filtered
  • Nuclease-free solution
  • Routinely used in DNA and RNA precipitation, elution of proteins and other uses in molecular biology applications
This Molecular Biology Grade sodium acetate solution ensures exceptional purity and quality, making it ideal for a wide range of laboratory applications, including nucleic acid purification, protein crystallization, and molecular assays. With its precisely formulated concentration and stringent quality control measures, CellPro™ Sodium Acetate provides researchers with reliable and reproducible results. Trust CellPro™ Sodium Acetate for your molecular biology experiments, and experience the confidence that comes with superior quality and performance.