4-Way Tube Rack
Supplier: Polycarbin
R04-01-01
77569-834CS 71.54 USD
77569-834
4-Way Tube Rack
Racks Tube Racks
The 4-Way tube rack accomodates 32×0.5 ml tubes, 32×1.5 to 2 ml tubes, 12×5 to 15 ml tubes, and 4×50 ml tubes.
  • Rack accomodates 0.5 to 50 ml tubes
  • Sturdy, polypropylene composition
  • Embossed numbers for organization purposes
  • Autoclavable, freezer safe
Polycarbin's 4-Way tube racks are perfect for sustainability-minded scientists engaging in benchtop work or cold storage. These compact, four-sided polypropylene racks are made from 100% post-consumer recycled content, yielding a tube rack with a significantly reduced carbon footprint.