CellPro™ Vacuum Filtration Flasks, PES Membrane
Supplier: Alkali Scientific
V05022 V05045 V100022 V100045 V15022 V15045 V25022 V25045 V50022 V50045
104027-496EA 126.14 USD
104027-496 104028-786 104027-594 104026-728 104027-228 104028-706 104027-050 104028-170 104028-382 104027-292
CellPro™ Vacuum Filtration Flasks, PES Membrane
Flasks Filter Flasks
Enhance your laboratory filtration processes with CellPro™ vacuum filtration flasks featuring PES membrane, ensuring efficient filtration.
  • Individually wrapped/Sterile
  • Gamma sterilized, pyrogen free
  • Engraved graduation marks
  • Lowest protein binding
  • Recommended use is >−60 kPa (kilopascal)
  • Maximum use is >−90 kPa (kilopascal)
Introducing CellPro™ vacuum filtration flasks with PES membrane, designed to streamline your filtration processes with precision and reliability. Available in 0.22 µm and 0.45 µm pore sizes, these filtration flasks feature a screw-on funnel equipped with a lid for secure handling. The filtration section boasts a graduated connection compatible with various hoses, facilitating seamless integration into your workflow. The receiving bottle is thoughtfully crafted with indentations for effortless gripping, along with a separate cap for added convenience. Both the funnel and receiver are adorned with engraved graduation marks, ensuring precise measurements. Moreover, these flasks undergo gamma sterilization and are pyrogen-free, guaranteeing the integrity of your filtrates. Upgrade your filtration setup with CellPro™ vacuum filtration flasks for unparalleled performance and peace of mind.