myGel™ Mini Electrophoresis All In One DNA Gel System
Supplier: Alkali Scientific
myGel™
ES1101
104028-724EA 693.83 USD
104028-724
myGel™ Mini Electrophoresis All In One DNA Gel System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Streamline DNA gel electrophoresis with the myGel™ Mini - electrophoresis all in one DNA gel system, designed for efficient analysis.
  • Gel box and safety lid
  • Power supply
  • Two 10.6×6 cm gel trays
  • Four 5×6 cm gel trays
  • Casting stands for the above trays
  • Two double sided combs for each gel size
The myGel™ horizontal electrophoresis system includes all the equipment you need to get up and running - gel tank, power supply and two complete casting sets. Ideal for personal use, small laboratories, or the classroom, this gel box/power supply combination is compact and easy to use.