CellPro™ EDTA, Molecular Biology Grade
Supplier: Alkali Scientific
DB0251 DB0254 DB0259 DB0316 DB0316-CS DB0276
104027-328EA 36.91 USD
104027-328 104028-884 104027-386 104028-104 104027-036 104028-392
CellPro™ EDTA, Molecular Biology Grade
Buffers Cloning Buffers
Maintain chelation activity with CellPro™ EDTA, meeting molecular biology grade standards for purity and reliability.
  • Sterile-filtered solution
  • For biochemistry or molecular biology applications requiring a chelator of divalent metal ions
CellPro™ EDTA, molecular biology grade, a high-quality solution designed for molecular biology applications. This molecular biology-grade EDTA offers exceptional purity and reliability, ensuring optimal performance in a variety of laboratory procedures. With its sterile formulation and molecular biology-grade quality, CellPro™ EDTA guarantees the preservation of sample integrity and the prevention of nucleic acid degradation.