CellPro™ TBE (Tris-borate EDTA), Sterile, Filtered
Supplier: Alkali Scientific
DB0216 DB0216-20L DB0216-4L DB0333 DB0333-CS DB2216-20L PB0290
104028-402EA 52.29 USD
104028-402 104027-042 104028-206 104028-606 104027-914 104027-996 104028-686
CellPro™ TBE (Tris-borate EDTA), Sterile, Filtered
Buffers Electrophoresis Buffers Running Buffers
Maintain nucleic acid integrity with sterile and filtered CellPro™ TBE (Tris-borate EDTA) buffer.
  • Compatible with horizontal agarose and vertical polyacrylamide gels
  • Use with nondenaturing and denaturing (with urea) gels
  • Better than TAE for long runs due to higher buffering capacity and less heating
  • Made with 18 Ω water
CellPro™ TBE (Tris-borate EDTA) solution offers a sterile and filtered formulation of this essential buffer for molecular biology and electrophoresis applications. With its meticulous preparation, this solution ensures purity and consistency, meeting the rigorous demands of scientific research and experimentation. The sterile nature of the solution minimizes the risk of contamination, making it ideal for various molecular biology techniques, including DNA and RNA agarose gel electrophoresis. Additionally, the filtered formulation guarantees clarity and reliability in experimental results, providing researchers with confidence in their findings. Whether used for nucleic acid separation, DNA fragment analysis, or other molecular biology procedures, CellPro™ TBE solution delivers consistent performance and reproducible results, supporting the success of laboratory workflows.