CellPro™ Culture Tubes
Supplier: Alkali Scientific
CT5305 CT5320 CT5225 CT5235 CT5250 CT5260 CT5300 CT5301
104027-300EA 181.52 USD
104027-300 104028-932 104028-754 104028-658 104027-566 104026-856 104028-808 104028-654
CellPro™ Culture Tubes
Tubes Culture Tubes
Cultivate your cells with precision in CellPro™ culture tubes, designed for optimal growth conditions.
  • Sterile, RNase, DNase, pyrogen and cytotoxin free
  • Polypropylene
  • Dual-position plug caps
  • Compatible with all standard equipment
  • Polypropylene tubes - centrifuged up to 4000 ×g
Disposable sterile culture tubes are made of high quality polypropylene (PP) they will withstand temperatures from −190 to +120 ˚C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polypropylene tubes can be centrifuged at up to 4000 ×g.