Ultra Low Attachment Dish with External Grip Edge
Supplier: Alkali Scientific
CFD100 CFD35 CFD60
104028-424EA 244.6 USD
104028-424 104026-594 104028-700
Ultra Low Attachment Dish with External Grip Edge
Culture Dishes Cell Culture Dishes
Ensure minimal cell adhesion with the ultra low attachment dish featuring an external grip edge for easy handling.
  • Ultra low cell adhesion
  • Comprised of a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation
  • Use when culturing stem cells and regenerative cell therapy
  • Promotes growth of suspension cell lines - both established and primary cells
  • Ideal environment to help nurture embryoid body formation from ES cells
  • Product shelf life: 3 years at room temperature storage (25 °C)
Ultra low attachment (ULA) cell tissue culture dishes offer a solution for cell culture requiring minimal cell adhesion. Featuring a covalently bound hydrogel layer, these dishes minimize cell attachment, protein absorption, and cellular activation. They are particularly suitable for culturing stem cells and regenerative cell therapy applications. These dishes promote the growth of suspension cell lines, including both established and primary cells, providing an ideal environment for nurturing embryoid body formation from ES cells. With a flat bottom design, they offer stability during cell culture procedures. These ULA dishes have a product shelf life of 3 years when stored at room temperature (25 ℃), ensuring long-term usability and reliability for research needs.