Conical Centrifuge Tubes, Sterile
Supplier: Alkali Scientific
C8225 C8226 C8227 C8250 C8252
104028-220EA 549.22 USD
104028-220 104026-732 104027-612 104027-134 104026-998
Conical Centrifuge Tubes, Sterile
Tubes Centrifuge Tubes
These 250 and 500 ml conical centrifuge tube can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences and clinical experiments and can meet the requirements of biological analysis.
  • The sealing structure design of the cover provides a two-way security to guarantee seal ability
  • The thickened design of the conical bottom increases the maximum centrifugal limit
  • Max. centrifugal limit 7000 ×g
  • Tolerable temperature −80 to +120 °C
  • The clear scale is convenient for data reading
  • No pyrogens, sterile, no DNA/RNA
  • Electron beam sterilization, SAL= 10⁻⁶