QwikSnap™ MicroTubes, Low Binding and Low Adhesion
Supplier: Alkali Scientific
C3019 C3021
104026-882EA 235.37 USD
104026-882 104027-728
QwikSnap™ MicroTubes, Low Binding and Low Adhesion
Tubes Microtubes
Ensure minimal sample loss with Qwik-Snap MicroTubes, featuring low binding and low adhesion properties.
  • Centrifugal Speed: 29000 ×g
  • Withstands boiling up to 121 °C and freezing down to -80 °C
  • Snaps into place for a secure seal
  • 100% polypropylene
  • RNase/DNase free
  • Clear, non-binding surface, graduated
  • Certified RNAse, DNAse and pyrogen free
  • Sterile options (SAL 10-6) and assorted colors available
QwikSnap™ MicroTubes, low binding and low adhesion are made of premium polypropylene plastic which means a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. The 0.6 ml tubes are packaged in bags of 1000 and the 1.7 ml and 2 ml tubes come 500 per pack. Available in 1.7 ml (graduated to 1.5 ml), 2.0 ml, and 0.6 ml sizes, the 1.7 ml and 0.6 ml tubes are also available in assorted colors. Capable of withstanding boiling temperatures up to 121 °C and freezing down to −80 °C, the Qwik-Snap micro centrifuge tubes are the perfect choice for research.