Cell Scrapers
Supplier: Alkali Scientific
C30020 C30040
104027-456EA 69.22 USD
104027-456 104028-798
Cell Scrapers
Cell Scrapers
Effortlessly harvest cells with cell scrapers.
  • Rotatable blades
  • Individually packaged
  • Pyrogen free
  • Gamma sterilized
The cell scraper is a specialized tool designed for the efficient collection of adherent cells from culture vessels. Available in a case of 100, these cell scrapers are ideal for cell culture and tissue culture applications. They feature a comfortable handle and a blade that allows easy and precise scraping of cells from the surface of culture dishes or flasks. With their sterile and disposable design, Cell Scraper offers convenience and reliability in cell harvesting procedures, making them essential for laboratory work involving cell culture studies.