SpectraTube™ Centrifuge Tubes, 500 Per Case
Supplier: Alkali Scientific
C2715 C2716 C2750 C2751
104027-784EA 167.68 USD
104027-784 104027-972 104026-986 104028-166
SpectraTube™ Centrifuge Tubes, 500 Per Case
Tubes Centrifuge Tubes
Ensure reliable sample processing with SpectraTube™ centrifuge tubes, available in a case of 500.
  • Colored caps for easy sample identification
  • Sterile, RNase, DNase and Pyrogen free
  • Made of premium polypropylene, with exceptionally uniform wall thickness for clarity and centrifugal strength
  • Leak proof
  • Quantity: 500 per case
ASI's new SpectraTube™ is a new twist on a staple in the life science laboratory. We've taken the proven design of the 15 and 50 ml conical tube and configured them conveniently in a foam rack with five different colored caps to make sample identification a breeze. These tubes are perfect for any lab conducting multiple studies, or any experiments requiring multiple sample types or solutions. All five cap varieties are flat, brightly colored, and frosted to make them easy to write on. These conical 15 and 50 ml centrifuge tubes are made of premium polypropylene, with exceptionally uniform wall thickness for clarity and centrifugal strength. Tubes and caps may be used for sample storage at temperatures as low as −45 ˚C. The maximum centrifugal forces are 12,000 ×g for the 15 ml tubes and 15,000 ×g for 50 ml version.* Tubes and caps are suitable for stem cell research and certified sterile, RNase, DNase, and Pyrogen free. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.