96 Well Cell Culture Plates- White, Sterile
Supplier: Alkali Scientific
TPW96 TPW9096
104027-340EA 87.68 USD
104027-340 104028-298
96 Well Cell Culture Plates- White, Sterile
Microplates Cell Culture Plates
Facilitate cell culture experiments with 96 well cell culture plates- white, supplied sterile in cases of 50 for reliable cell growth and experimentation.
  • HTS (High-Throughput Screening) compatible
  • Suitable for fluorescence assay, luminescence assay
  • TC Treated for cell culture and adherent cell types
  • Designed to prevent cross contamination
  • Effective gas exchange lid inner design
  • Alphanumeric labeling, homogeneous cell culture
  • Flat bottom, USP guideline compliant (USP class Ⅵ tested)
  • ANSI guideline compliant
Elevate the precision of your luminescence assays with our 96-well white cell culture plates. Designed for optimal reflection and minimal cross-talk, these plates feature a uniform white color throughout the entire body and flat surface. Each plate is sterile and comes with a compatible, low-evaporation lid to maintain sample integrity. With 50 plates per case, you'll have an ample supply for your research needs.