Cell Culture Plates with Lids, PS, 96 well, Black Wells and Flat Bottom, Treated
Supplier: Alkali Scientific
TPB9096 TPB96
104027-552EA 126.14 USD
104027-552 104027-810
Cell Culture Plates with Lids, PS, 96 well, Black Wells and Flat Bottom, Treated
Microplates Cell Culture Plates
Ensure reliable cell growth in the cell culture plates, featuring polystyrene construction with 96 wells, black wells, and flat bottom design, treated for cell culture, and supplied sterile with lids, packaged in a case of 50.
  • Black sidewalls/black bottom
  • Bottom material: PS
  • Designed to prevent well to well cross contamination
  • Effective gas exchange lid inner design
  • Alphanumeric labeling
  • High throughput screening compatible
  • Suitable for fluorescence and luminescence assays
  • Non treated for suspension cultures
  • Designed to prevent cross contamination
Experience unparalleled performance in your cell culture experiments with the sterile 96-well cell culture plates. These plates, constructed from premium polystyrene, feature a design with black wells and a flat bottom, enhancing optical clarity and facilitating imaging and fluorescent detection. Engineered with a specialized treatment, these plates promote superior cell adhesion, ensuring optimal cell growth and viability. Each plate comes with a secure lid, preserving sample integrity and maintaining sterility throughout your experiments.