Cell Culture Plates with Lids, PS, 96-Well, Black Well and Clear Flat Bottom, Sterile, Treated
Supplier: Alkali Scientific
TPB1096
104027-200EA 361.52 USD
104027-200
Cell Culture Plates with Lids, PS, 96-Well, Black Well and Clear Flat Bottom, Sterile, Treated
Microplates Cell Culture Plates
Perform assays with precision using the cell culture plates, made of polystyrene with 96 wells, black well, and clear flat bottom design, treated for cell culture, and supplied sterile with lids, available in a case of 25.
  • Black wall and topside (PS)/clear bottom
  • Bottom material: Optically clear PS
  • Designed to prevent cross contamination
  • Effective gas exchange lid inner design
  • Alphanumeric labeling
  • Provided with matching low evaporation lid
Experience unparalleled versatility and reliability with sterile 96-well cell culture plates. This premium plate combines the durability of polystyrene construction with the convenience of a black well and clear flat bottom design. Each plate is meticulously treated to ensure optimal cell adhesion and growth, making it ideal for a wide range of cell culture applications. The inclusion of a secure lid provides added protection and maintains sterility throughout your experiments.