96-Well Deep Well Plates, 2.2 ml, V Bottom, Square Well
Supplier: Alkali Scientific
TL455
104027-582EA 272.29 USD
104027-582
96-Well Deep Well Plates, 2.2 ml, V Bottom, Square Well
Microplates Deep Well Microplates
Perform high-throughput extractions with 2.2 ml 96-well deep well plate, featuring a V bottom, square well design suitable for KingFisher extraction platform, and certified DNase and pyrogen free in a pack of 50.
  • High chemical stability
  • Temperature range: −80 to +120 °C
  • Autoclavable
  • Centrifugal strength up to 3000 ×g
  • Compatible with multi-channel pipette and automatic equipment
  • Optimized bottom design to minimize residual volume
  • Clear alphanumeric codes for easy identification
  • Conform to SBS/ANSI
This 2.2 ml 96-well deep well plate, specifically designed for use with the KingFisher extraction platform. Featuring a V-bottom design and square wells, these plates ensure efficient sample recovery and extraction. Manufactured to be DNase and pyrogen-free, they provide reliable performance and maintain sample integrity, crucial for downstream applications. With a capacity of 2.2 ml per well, these plates offer ample space for sample processing, making them ideal for high-throughput applications in molecular biology, genomics, and proteomics research.
Specific 96 well deep plate used in nucleic acid extraction and its stackable, reliable tight sealing,non-pyrogenic, DNase/RNase free.