Premium Glass Bottom Cell Culture Dish
Supplier: Alkali Scientific
TC801002 TC801004 TC801006
104028-600EA 693.83 USD
104028-600 104026-576 104027-794
Premium Glass Bottom Cell Culture Dish
Culture Dishes Cell Culture Dishes
Facilitate live-cell imaging with the premium glass bottom cell culture dish, providing optimal optical clarity for microscopy. Use this dish with confocal microscopes, high resolution microscopes, differential interference contrast microscopes, polarized light microscopes, and phase contrast microscopes for cell observation.
  • Can be used for live cells observation
  • Special bottom design for easy grip
  • Imported optical grade cover glass, cover glass thickness: 0.19 to 0.22 mm
  • Round cover glass insert for good appearance
  • Medical adhesive to guarantee non-cytotoxicity
These premium glass bottom cell culture dishes are designed to meet the demands of advanced cell imaging and analysis techniques. Featuring a high-quality glass bottom, these dishes offer superior optical clarity, enabling high-resolution imaging of cellular structures and dynamics in live-cell experiments.