Cell Lifter, Double Blade
Supplier: Alkali Scientific
TC7023
104027-956EA 47.68 USD
104027-956
Cell Lifter, Double Blade
Cell Scrapers
Efficiently handle cell cultures with the cell lifter, equipped with a double blade featuring 23 and 12 mm edges and a 218 mm handle.
  • Useful for minimal harvesting of cells
  • Wide/narrow baldes
  • Individually packed
  • Pyrogen free
  • Sterile
It provides ergonomic support and ensures efficient and gentle detachment of adherent cells from culture vessels, minimizing cell damage and preserving cell viability.