CellPro™ PolyPestle Double-sided Pestle
Supplier: Alkali Scientific
CellPro™
MT4010
104028-380EA 50.75 USD
104028-380
CellPro™ PolyPestle Double-sided Pestle
Mortars and Pestles Pestles
Enhance laboratory workflows with the CellPro™ PolyPestle, a double-sided pestle designed for use with cell strainers and 1.5 ml microtubes.
  • Specifically designed for use with cell strainers and 1.5 ml microtubes
  • Sterile
  • Length: 5.75" (145 mm)
  • Individually wrapped
Streamline sample processing with the CellPro™ PolyPestle. This double-sided pestle is designed for efficient homogenization of samples in cell strainers and 1.5 ml microtubes. The ergonomic design ensures comfortable handling and effective sample disruption, making it ideal for a variety of laboratory applications. Trust in the reliability and performance of the CellPro™ PolyPestle to enhance the efficiency of sample preparation workflows.