CellPro™ SSC Buffer 20X, Molecular Biology Grade, Sterile Filtered
Supplier: Alkali Scientific
DS0324 DS0370-20L DS0370-4L DS0370-CS PB0285
104026-734EA 53.83 USD
104026-734 104026-822 104026-584 104027-942 104028-086
CellPro™ SSC Buffer 20X, Molecular Biology Grade, Sterile Filtered
Buffers Electrophoresis Buffers Hybridization Buffers
Prepare hybridization buffers with CellPro™ SSC buffer 20X, provided sterile filtered and meeting molecular biology grade standards.
  • For molecular biology
  • Liquid medium
  • 0.2 μm filtered
CellPro™ SSC buffer 20X stands as a cornerstone of excellence in molecular biology research. Crafted to meticulous standards of purity and sterility, this buffer offers researchers a reliable solution for nucleic acid hybridization experiments. With its sterile filtered formulation, CellPro™ SSC Buffer ensures consistent and reproducible results, empowering researchers in their molecular biology endeavors.