CellPro™ Culture Tubes
Supplier: Alkali Scientific
CT8832 C3095-7 CT8730 CT8830
104027-890EA 333.83 USD
104027-890 104027-762 104027-108 104028-508
CellPro™ Culture Tubes
Tubes Culture Tubes
Ensure consistent cell culture experiments with CellPro™ Culture tubes, providing reliable results every time.
  • Aerobic culturing at first click
  • Anaerobic after second click
  • Printed graduations
  • Flat, markable screw-cap
CellPro™ disposable sterile culture tubes are made of high quality polypropylene (PP) they will withstand temperatures from −190 to +120 °C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). These polypropylene tubes can be centrifuged at up to 4000 ×g.