CellPro™ LB and Carbenicillin
Supplier: Alkali Scientific
CellPro™
DC0081 DC0082 DC4470 DC0085 DC4480 DC0087 DC0084 DC0086 DC0088 DC0091 DS0171 DC0089 DC0083
104028-864EA 56.91 USD
104028-864 104028-152 104028-144 104028-002 104029-078 104027-018 104027-864 104027-978 104027-186 104027-030 104027-382 104027-542 104027-124
CellPro™ LB and Carbenicillin
Media for Microbiology Prepared Media for Microbiology
Facilitate bacterial culture with CellPro™ LB and Carbenicillin, an ideal medium for maintaining plasmids in transformed cells.
  • Elevate your selections with CellPro™'s blend of LB broth and carbenicillin
  • Ensure efficient bacterial growth and selection for your experiments
  • Trust in consistent performance for reliable research outcomes
CellPro™ LB and Carbenicillin is a specialized culture medium containing carbenicillin, a β-lactam antibiotic used to select for bacterial strains carrying carbenicillin resistance genes. This LB agar variant is co mmonly employed in molecular biology research for the cultivation and maintenance of recombinant bacterial cells harboring carbenicillin resistance plasmids. By incorporating carbenicillin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of non-resistant contaminants.