BrightStar™ Duration HRP Chemiluminescent Substrates
Supplier: Alkali Scientific
XR91 XR92
104027-618EA 70.75 USD
104027-618 104026-868
BrightStar™ Duration HRP Chemiluminescent Substrates
Electrophoresis Reagents Western Blotting Reagents
Achieve sensitive detection in chemiluminescence assays with the BrightStar™ Duration HRP Chemiluminescent Substrate, designed for accurate and efficient protein detection.
  • Suitable detection for picogram levels of protein
  • Sensitive and long lasting luminol-based
The BrightStar™ Duration HRP Chemiluminescent Substrate is a sensitive and long lasting luminol-based chemiluminescent two solution kit that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as either a two component kit system for e×tended shelf-life, or as a single component mix system for added convenience. Visualization may be carried out through x-ray film or digital imaging system utilizing a CCD camera.