CellPro™ Vacuum Filtration Flasks, PVDF Membrane, Sterile
Supplier: Alkali Scientific
VP05045 VP100022 VP100045 VP15022 VP15045 VP25022 VP25045 VP50022 VP50045
104028-384EA 156.91 USD
104028-384 104027-040 104027-608 104029-100 104026-794 104027-934 104026-952 104028-824 104029-036
CellPro™ Vacuum Filtration Flasks, PVDF Membrane, Sterile
Flasks Filter Flasks
Improve your filtration processes with CellPro™ vacuum filtration flasks featuring PVDF membrane, individually wrapped and sterile, available in a case of 12.
  • Individually wrapped
  • Ga mma sterilized, pyrogen free
  • Engraved graduation marks
  • Low protein binding
CellPro vacuum filtration flasks are equipped with either 0.22 or 0.45 µm PVDF membrane filters. Featuring a screw-on funnel with a lid, a graduated connection in the filtration section to acco mmodate multiple hoses, and a receiving bottle with indentations for easy gripping and a separate cap, these flasks are designed for convenient use. Engraved graduation marks on both the funnel and receiver ensure accuracy, and the entire assembly is ga mma sterilized and pyrogen free. Packaged in cases of 12, each flask is individually wrapped for hygiene and further assurance of sterility. With low protein binding properties, these flasks are suitable for a variety of filtration tasks. The 1000 ml vacuum filter flasks are ideal for purifying tissue culture media and other liquid samples. Additionally, CellPro bottle top filters are available separately for users who do not require receiving bottles.